missing translation for 'onlineSavingsMsg'
Learn More

Cesium chloride, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11320328
250 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Cesium chloride is widely utilized as a precursor for the source of cesium ions. It finds application in analytical chemistry and acts as a reagent to identify inorganic ions like aluminum, gallium and chromium by the color and morphology of the precipitate. Its radioisotopes are employed in nuclear medicine applications such as treatment of cancer and diagnosis of myocardial infarction. Occasionally, it can act as a phase transfer catalyst in selected organic reactions. It is applied in the manufacturing of electrically conducting glasses and screens of cathode ray tubes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7647-17-8
Molecular Formula ClCs
Molecular Weight (g/mol) 168.36
MDL Number MFCD00010955
InChI Key AIYUHDOJVYHVIT-UHFFFAOYSA-M
Synonym cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chlorideShow MoreShow Less
PubChem CID 24293
ChEBI CHEBI:63039
SMILES [Cl-].[Cs+]

Specifications

Boiling Point 1303°C
Melting Point 646°C
Color White
Physical Form Beads
Quantity 250 g
Assay Percent Range 99.99% (Metals basis)
Solubility Information Soluble in water and alcohol. Insoluble in acetone.
IUPAC Name cesium;chloride
Formula Weight 168.36
Odor Odorless
Refractive Index 1.6418
Sensitivity Hygroscopic
Density 3.99
Chemical Name or Material Cesium chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU07K030 Certificate TypeCertificate of Analysis Date22/08/2024 Catalog Number043720.09,043720.30,043720.18
Lot NumberP07K008 Certificate TypeCertificate of Analysis Date13/03/2024 Catalog Number043720.09,043720.30,043720.18
Lot NumberM02K036 Certificate TypeCertificate of Analysis Date10/01/2024 Catalog Number043720.09,043720.30,043720.18
Lot NumberY09G029 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number043720.09,043720.30,043720.18
Lot NumberZ02H036 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number043720.09,043720.30,043720.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cesium chloride, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.