missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cesium Chloride, Ultrapure Bioreagent, J.T. Baker™
High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography
£278.00 - £1920.00
Chemical Identifiers
CAS | 7647-17-8 |
---|---|
Molecular Formula | ClCs |
Molecular Weight (g/mol) | 168.36 |
MDL Number | MFCD00010955 |
InChI Key | AIYUHDOJVYHVIT-UHFFFAOYSA-M |
Synonym | cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride |
PubChem CID | 24293 |
ChEBI | CHEBI:63039 |
IUPAC Name | cesium;chloride |
SMILES | [Cl-].[Cs+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15538224
|
Avantor J.T.Baker
4042-04 |
100 g | Poly bottle |
£278.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15528224
|
Avantor J.T.Baker
4042-02 |
1 kg | Poly Bottle |
£1920.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- For Density Gradient Centrifugation; For Laboratory,Research,or Manufacturing Use
Chemical Identifiers
7647-17-8 | |
168.36 | |
AIYUHDOJVYHVIT-UHFFFAOYSA-M | |
24293 | |
cesium;chloride |
ClCs | |
MFCD00010955 | |
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride | |
CHEBI:63039 | |
[Cl-].[Cs+] |
Specifications
7647-17-8 | |
MFCD00010955 | |
AIYUHDOJVYHVIT-UHFFFAOYSA-M | |
cesium;chloride | |
24293 | |
168.36 | |
1L = 3.99kg |
ClCs | |
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride | |
[Cl-].[Cs+] | |
168.36 | |
CHEBI:63039 | |
Ultrapure Bioreagent | |
Cesium Chloride |