missing translation for 'onlineSavingsMsg'
Learn More
Cesium Chloride (White Crystalline Powder/Molecular Biology), Fisher BioReagents Product Code.: 10734344

Cesium Chloride (White Crystalline Powder/Molecular Biology), Fisher BioReagents

Product Code. 10734344
100 g, Poly Bottle
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy

Product Code. 10734344

Brand: Fisher BioReagents BP210100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
This Molecular Biology-grade cesium chloride is ideal for UV analysis of nucleic acids during density-gradient centrifugation and for the isolation of plasmid DNA, phage DNA, virus particles, and RNA.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-17-8
Molecular Formula ClCs
Molecular Weight (g/mol) 168.36
MDL Number MFCD00010955
InChI Key AIYUHDOJVYHVIT-UHFFFAOYSA-M
Synonym cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
PubChem CID 24293
ChEBI CHEBI:63039
IUPAC Name caesium(1+) chloride
SMILES [Cl-].[Cs+]

Specifications

Boiling Point 1290°C
Melting Point 646°C
Color White
pH 5 to 9
Physical Form Crystalline Powder
Quantity 100 g
Assay Percent Range ≥99.999 %
Merck Index 15, 2010
Formula Weight 168.36
Percent Purity ≥99.999%
Grade DNase-, RNase- and Protease-Free
Packaging Poly Bottle
Refractive Index 1.367 to 1.371 (at 25°C)
Chemical Name or Material Cesium Chloride
Show More Show Less
compliance-icons
Product Identifier
  • Cesium chloride
Signal Word
  • Warning
Hazard Category
  • Reproductive toxicity Category 2
Hazard Statement
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cesium Chloride (White Crystalline Powder/Molecular Biology), Fisher BioReagents >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.