missing translation for 'onlineSavingsMsg'
Learn More
Cesium fluoride, 99.9%, extra pure, Thermo Scientific Chemicals Product Code.: 10392823

Cesium fluoride, 99.9%, extra pure, Thermo Scientific Chemicals

Product Code. 10392823
5 g, Plastic bottle
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10392823

Brand: Thermo Scientific Acros 315910050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13400-13-0
Molecular Formula CsF
Molecular Weight (g/mol) 151.90
MDL Number MFCD00010960
InChI Key XJHCXCQVJFPJIK-UHFFFAOYSA-M
Synonym cesium fluoride, caesium fluoride, cesium monofluoride, dicesium difluoride, tricesium trifluoride, unii-t76a371hjr, caesium 1+ ion fluoride, caesiumfluoride, cesiumfluoride
PubChem CID 25953
SMILES [F-].[Cs+]

Specifications

Boiling Point 1251.0°C
Melting Point 682.0°C
Color White
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.9%
Linear Formula CsF
Fieser 01,121; 06,100; 07,57; 08,81; 09,100; 10,81; 11,115; 12,108; 13,68; 14,79; 15,75; 16,69; 17,68
Merck Index 15, 2011
Solubility Information Solubility in water: 369g/100mL (18°C). Other solubilities: soluble in methanol, insoluble in dioxane and pyridine
IUPAC Name caesium(1+) fluoride
Formula Weight 151.91
Percent Purity 99.9%
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Cesium fluoride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0443011 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number315910050,315911000,315910250
Lot NumberA0461137 Certificate TypeCertificate of Analysis Date18/03/2024 Catalog Number315910050,315911000,315910250
Lot NumberA0395001 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number315910050,315911000,315910250
Lot NumberA0443011 Certificate TypeCertificate of Analysis Date10/06/2022 Catalog Number315910050,315911000,315910250
Lot NumberA0431073 Certificate TypeCertificate of Analysis Date08/06/2021 Catalog Number315910050,315911000,315910250
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cesium fluoride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
Hazard Statement
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H361-Suspected of damaging fertility or the unborn child.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cesium fluoride, 99.9%, extra pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.