missing translation for 'onlineSavingsMsg'
Learn More
Cesium fluoride, 99.9% (metals basis), Thermo Scientific Chemicals Product Code.: 11356717

Cesium fluoride, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11356717
100 g
Click to view available options
Quantity:
2.5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11356717

Brand: Thermo Scientific Alfa Aesar 010019.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for desilylation reactions in organic synthesis. Useful base in organic chemistry, due to its low nucleophilicity. Owing to its high ionic character, it is a very reactive source of fluoride ion in organic synthesis. Reagent of choice when reactive naked fluoride is required. Effective catalyst in promoting Knoevenegal condensation. Single crystals of CsF are useful as the windows of cells for infrared spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13400-13-0
Molecular Formula CsF
Molecular Weight (g/mol) 151.90
MDL Number MFCD00010960
InChI Key XJHCXCQVJFPJIK-UHFFFAOYSA-M
Synonym cesium fluoride, caesium fluoride, cesium monofluoride, dicesium difluoride, tricesium trifluoride, unii-t76a371hjr, caesium 1+ ion fluoride, caesiumfluoride, cesiumfluoride
PubChem CID 25953
SMILES [F-].[Cs+]

Specifications

Boiling Point 1251°C
Melting Point 682°C
Color White
Physical Form Lump/Powder
Quantity 100 g
Assay Percent Range 99.9% (Metals basis)
Solubility Information Soluble in water. Insoluble in dioxane and pyridine.
IUPAC Name cesium;fluoride
Formula Weight 151.9
Sensitivity Hygroscopic
Chemical Name or Material Cesium fluoride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cesium fluoride, 99.9% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.