missing translation for 'onlineSavingsMsg'
Learn More
Cesium hydroxide monohydrate, 99.95%, (trace metal basis), for analysis, Thermo Scientific Chemicals Product Code.: 10761081

Cesium hydroxide monohydrate, 99.95%, (trace metal basis), for analysis, Thermo Scientific Chemicals

Product Code. 10761081
250 g, Plastic bottle
Click to view available options
Quantity:
50 g
250 g
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10761081

Brand: Thermo Scientific Acros 189522500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35103-79-8
Molecular Formula CsH3O2
Molecular Weight (g/mol) 167.93
MDL Number MFCD00149664
InChI Key ABSOMGPQFXJESQ-UHFFFAOYSA-M
Synonym cesium hydroxide monohydrate, cesium hydroxide hydrate, cesium hydroxide, monohydrate, cesium hydroxide-hydrate, cesium hydroxide, hydrate, ksc493c5l, caesium 1+ ion hydrate hydroxide, cesium hydroxide,monohydrate 9ciShow MoreShow Less
PubChem CID 23679066
SMILES O.[OH-].[Cs+]

Specifications

Melting Point 205.0°C to 208.0°C
Color Blue-Red to Green
Physical Form Powder
Quantity 250 g
Assay Percent Range 99.95%
Linear Formula CsOH·H2O
Merck Index 10, 1970
Solubility Information Solubility in water: soluble with exothermic reaction. Other solubilities: soluble in alcohol (exothermic)
IUPAC Name cesium;hydroxide;hydrate
Formula Weight 167.93
Percent Purity 99.95%
Grade Trace Metal Basis
Packaging Plastic bottle
Chemical Name or Material Cesium hydroxide monohydrate, 99.5%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465807 Certificate TypeCertificate of Analysis Date10/09/2024 Catalog Number189520500,189522500
Lot NumberA0451523 Certificate TypeCertificate of Analysis Date31/03/2024 Catalog Number189520500,189522500
Lot NumberA0393541 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number189520500,189522500
Lot NumberA0397591 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number189520500,189522500
Lot NumberA0432772 Certificate TypeCertificate of Analysis Date18/08/2022 Catalog Number189520500,189522500
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cesium hydroxide monohydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H361-Suspected of damaging fertility or the unborn child.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cesium hydroxide monohydrate, 99.95%, (trace metal basis), for analysis, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.