missing translation for 'onlineSavingsMsg'
Learn More
Cesium iodide, 99.9%, for analysis, Thermo Scientific Chemicals Product Code.: 10554131

Cesium iodide, 99.9%, for analysis, Thermo Scientific Chemicals

Product Code. 10554131
25 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10554131

Brand: Thermo Scientific Acros 192050250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-17-5
Molecular Formula CsI
Molecular Weight (g/mol) 259.81
MDL Number MFCD00010962
InChI Key XQPRBTXUXXVTKB-UHFFFAOYSA-M
Synonym cesium iodide, caesium iodide, cesium monoiodide, dicesium diiodide, tricesium triiodide, cesiumiodide, cesium iodide cs2i2, cesium iodide cs3i3, unii-u1p3gvc56l
PubChem CID 24601
IUPAC Name cesium;iodide
SMILES [I-].[Cs+]

Specifications

Boiling Point 1280.0°C
Melting Point 626.0°C
Color White
Physical Form Crystalline Powder
Quantity 25 g
Assay Percent Range 99.9%
Linear Formula CsI
Merck Index 15, 2013
Solubility Information Solubility in water: 740g/L (20°C). Other solubilities: soluble in ethanol, slightly soluble in methanol, practically insoluble in acetone
Formula Weight 259.8
Percent Purity 99.9%
Grade Analytical
Packaging Glass bottle
Chemical Name or Material Cesium iodide, 99.9%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465250 Certificate TypeCertificate of Analysis Date19/08/2024 Catalog Number192051000,192050250
Lot NumberA0457329 Certificate TypeCertificate of Analysis Date14/06/2024 Catalog Number192051000,192050250
Lot NumberA0455273 Certificate TypeCertificate of Analysis Date14/06/2024 Catalog Number192051000,192050250
Lot NumberA0393542 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number192051000,192050250
Lot NumberA0397415 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number192051000,192050250
compliance-icons compliance-icons
Product Identifier
  • Cesium iodide
Signal Word
  • Warning
Hazard Category
  • ACUTE AQUATIC Acute 1
  • Reproductive toxicity Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cesium iodide, 99.9%, for analysis, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.