missing translation for 'onlineSavingsMsg'
Learn More

Cesium metavanadate, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11323608
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14644-55-4
Molecular Formula CsO3V
Molecular Weight (g/mol) 231.844
MDL Number MFCD00049812
InChI Key SZIZYHNGMIYCQE-UHFFFAOYSA-N
Synonym Cesium vanadate, meta
PubChem CID 23689378
IUPAC Name cesium;oxido(dioxo)vanadium
SMILES [O-][V](=O)=O.[Cs+]

Specifications

Color White to Yellow
Physical Form Powder
Quantity 25 g
UN Number UN3288
Solubility Information Slightly soluble in water.
Formula Weight 231.84
Percent Purity 99.9%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Cesium metavanadate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'