missing translation for 'onlineSavingsMsg'
Learn More

Cesium methanesulfonate, 98%, Thermo Scientific Chemicals

£56.78 - £194.97

Chemical Identifiers

CAS 2550-61-0
Molecular Formula CH3CsO3S
Molecular Weight (g/mol) 228
InChI Key DJKJXRLREATOMF-UHFFFAOYSA-M
Synonym cesium methanesulfonate, csmes, csmsf, methanesulfonic acid cesium salt, caesium 1+ ion mesylate, cesium methanesulfonate used in patch clamp techniques, cesium methanesulfonate, used in patch clamp techniques
PubChem CID 5148066
IUPAC Name cesium;methanesulfonate
SMILES CS(=O)(=O)[O-].[Cs+]
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
11905891
View Documents
Thermo Scientific Acros
431580050
5 g Glass bottle
£56.78
5g
Please to purchase this item. Need a web account? Register with us today!
11935891
View Documents
Thermo Scientific Acros
431580250
25 g Glass bottle
£194.97
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2550-61-0
228
cesium methanesulfonate, csmes, csmsf, methanesulfonic acid cesium salt, caesium 1+ ion mesylate, cesium methanesulfonate used in patch clamp techniques, cesium methanesulfonate, used in patch clamp techniques
cesium;methanesulfonate
CH3CsO3S
DJKJXRLREATOMF-UHFFFAOYSA-M
5148066
CS(=O)(=O)[O-].[Cs+]

Specifications

262.0°C to 264.0°C
White
98%
cesium methanesulfonate, csmes, csmsf, methanesulfonic acid cesium salt, caesium 1+ ion mesylate, cesium methanesulfonate used in patch clamp techniques, cesium methanesulfonate, used in patch clamp techniques
DJKJXRLREATOMF-UHFFFAOYSA-M
cesium;methanesulfonate
5148066
98%
Cesium methanesulfonate
2550-61-0
Powder
CH3CsO3S
(5% in water) Clear colorless
CS(=O)(=O)[O-].[Cs+]
228
228
Authentic
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

Product Identifier
  • Cesium methanesulfonate

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only