missing translation for 'onlineSavingsMsg'
Learn More

Cesium solution for Ion Chromatography 1g/L H2O matrix, Fisher chemical™

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

£46.55 - £96.60

Chemical Identifiers

CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
12972095
View Documents Promotion Details
Fisher Chemical
A 010 100
100 mL Bottle
£46.55
100mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12982095
View Documents Promotion Details
Fisher Chemical
A 010 250
250 mL Bottle
£66.35
250mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12992095
View Documents Promotion Details
Fisher Chemical
A 010 500
500 mL Bottle
£96.60
500mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

7697-37-2
63.01
GRYLNZFGIOXLOG-UHFFFAOYSA-N
CHEBI:48107
O[N+]([O-])=O
HNO3
MFCD00011349
944
nitric acid

Specifications

1g/L
Single-component Standard Solution
MFCD00011349
O[N+]([O-])=O
63.01
CHEBI:48107
Liquid
7697-37-2
HNO3
GRYLNZFGIOXLOG-UHFFFAOYSA-N
nitric acid
944
H2O
Cesium
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 24 Months. (12 Months after opening)

SDS
Documents

Documents

Product Certifications