missing translation for 'onlineSavingsMsg'
Learn More
Cesium sulfate, 99.95% (metals basis), Thermo Scientific Chemicals Product Code.: 11306727

Cesium sulfate, 99.95% (metals basis), Thermo Scientific Chemicals

Product Code. 11306727
100 g
Click to view available options
Quantity:
100 g
This item is not returnable. View return policy

Product Code. 11306727

Brand: Thermo Scientific Alfa Aesar 010027.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cesium sulfate is used to prepare dense aqueous solutions, which finds application in isopycnic centrifugation especially for DNA purification and recovery. It acts as a catalyst for sulfur dioxide oxidation with vanadium or vanadium pentoxide. Further, it acts as a metal oxide catalyst enhancer for heterogeneous processes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10294-54-9
Molecular Formula Cs2O4S
Molecular Weight (g/mol) 361.87
MDL Number MFCD00010959
InChI Key FLJPGEWQYJVDPF-UHFFFAOYSA-L
Synonym cesium sulfate, dicesium sulfate, sulfuric acid, dicesium salt, caesium sulphate, caesium sulfate, unii-8d6r91cs62, dicaesium 1+ ion sulfate, cesiumsulfate, dicaesium 1+ sulfateShow MoreShow Less
PubChem CID 25137
SMILES [Cs+].[Cs+].[O-]S([O-])(=O)=O

Specifications

Melting Point 1010°C
Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.9% (Metal basis)
Solubility Information Very soluble in water. Slightly soluble in alcohol,acetone,pyridine.
IUPAC Name dicesium;sulfate
Formula Weight 361.87
Sensitivity Hygroscopic
Appearance White powder
Density 4.243
Chemical Name or Material Cesium sulfate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM24L006 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog Number010027.22,010027.14
Lot NumberU15K023 Certificate TypeCertificate of Analysis Date23/08/2024 Catalog Number010027.22,010027.14
Lot NumberQ23K061 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number010027.22,010027.14
Lot NumberN12K028 Certificate TypeCertificate of Analysis Date21/02/2024 Catalog Number010027.22,010027.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cesium sulfate, 99.95% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.