Learn More
Cesium sulfate, 99%, Thermo Scientific Chemicals
£29.00 - £356.00
Chemical Identifiers
CAS | 10294-54-9 |
---|---|
Molecular Formula | Cs2O4S |
Molecular Weight (g/mol) | 361.87 |
MDL Number | MFCD00010959 |
InChI Key | FLJPGEWQYJVDPF-UHFFFAOYSA-L |
Synonym | cesium sulfate, dicesium sulfate, sulfuric acid, dicesium salt, caesium sulphate, caesium sulfate, unii-8d6r91cs62, dicaesium 1+ ion sulfate, cesiumsulfate, dicaesium 1+ sulfate |
PubChem CID | 25137 |
IUPAC Name | dicesium;sulfate |
SMILES | [Cs+].[Cs+].[O-]S([O-])(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11402788
|
Thermo Scientific Alfa Aesar
A16767.14 |
25 g |
£29.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11412788
|
Thermo Scientific Alfa Aesar
A16767.22 |
100 g |
£84.50
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11422788
|
Thermo Scientific Alfa Aesar
A16767.36 |
500 g |
£356.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cesium sulfate is used to prepare dense aqueous solutions for use in isopycnic centrifugation. It is used as metal oxide catalyst enhancer for heterogeneous processes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCesium sulfate is used to prepare dense aqueous solutions for use in isopycnic centrifugation. It is used as metal oxide catalyst enhancer for heterogeneous processes.
Solubility
Soluble in water. (1820 g/L) at 20°C
Notes
Hygroscopic. Protect from humidity and water. Store under dry inert gas. Incompatible with water, reducing agents and oxidizing agents.
Chemical Identifiers
10294-54-9 | |
361.87 | |
FLJPGEWQYJVDPF-UHFFFAOYSA-L | |
25137 | |
[Cs+].[Cs+].[O-]S([O-])(=O)=O |
Cs2O4S | |
MFCD00010959 | |
cesium sulfate, dicesium sulfate, sulfuric acid, dicesium salt, caesium sulphate, caesium sulfate, unii-8d6r91cs62, dicaesium 1+ ion sulfate, cesiumsulfate, dicaesium 1+ sulfate | |
dicesium;sulfate |
Specifications
1,010°C | |
Cs2O4S | |
14,2016 | |
Soluble in water. (1820g/L) at 20°C | |
[Cs+].[Cs+].[O-]S([O-])(=O)=O | |
361.87 | |
361.87 | |
Odorless | |
4.24 |
10294-54-9 | |
MFCD00010959 | |
cesium sulfate, dicesium sulfate, sulfuric acid, dicesium salt, caesium sulphate, caesium sulfate, unii-8d6r91cs62, dicaesium 1+ ion sulfate, cesiumsulfate, dicaesium 1+ sulfate | |
FLJPGEWQYJVDPF-UHFFFAOYSA-L | |
dicesium;sulfate | |
25137 | |
99% | |
Hygroscopic | |
Cesium sulfate |
Safety and Handling
P201-P202-P260-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
H302-H361f-H373
missing translation for 'einecsNumber' : 233-662-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only