missing translation for 'onlineSavingsMsg'
Learn More

Cesium trifluoroacetate, Thermo Scientific Chemicals

Product Code. 11465450
Click to view available options
Quantity:
10 g
50 g

It is used in acid catalyzed reactions, especially those where an ester is cleaved in peptide synthesis. Used as an ion pairing agent in liquid chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21907-50-6
Molecular Formula C2CsF3O2
Molecular Weight (g/mol) 245.92
MDL Number MFCD00051304
InChI Key RJYSYRSELCQCSO-UHFFFAOYSA-M
Synonym cesium trifluoroacetate, caesio 2,2,2-trifluoroacetate, trifluoroacetic acid cesium salt, cesium trifluoroacetate solution, cstfa, caesium trifluoroacetate, cesium 2,2,2-trifluoroacetate, caesium 1+ trifluoroacetate, cesium 2,2,2-tris fluoranyl ethanoate, caesium 1+ ion trifluoroacetate
PubChem CID 23689558
IUPAC Name cesium;2,2,2-trifluoroacetate
SMILES [Cs+].[O-]C(=O)C(F)(F)F

Specifications

Melting Point 115°C
Physical Form Crystalline
Quantity 50 g
Solubility Information Soluble in water (2 M,clear,colorless) at 20°C. Soluble in organic solvents.
Formula Weight 245.92
Chemical Name or Material Cesium trifluoroacetate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.