Learn More
Tocris Bioscience™ CGP 71683 hydrochloride
Highly selective and potent non-peptide NPY Y5 receptor antagonist
Brand:  Tocris Bioscience™ 2199/50
668.42 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18005412
  • £846.00 / 50mg
Explore more special offers
Description
Description
CGP 71683 hydrochloride is an extremely selective, non-peptide NPY Y5 receptor antagonist. Displays > 1000-fold selectivity over Y1, Y2 and Y4 receptors; IC50 values are 1.4, 2765, 7187 and 5637 nM at cloned rat Y5, Y1, Y2 and Y4 receptors respectively. Potently inhibits NPY-induced food intake following i.p. administration in diabetic, free-feeding and fasted rats.
Specifications
Specifications
>99%
CGP 71683 hydrochloride
CGP 71683 hydrochloride
50 mg
512.07