Learn More
CHAC1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA542212
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: TPQNPGYLGP APEEAIATQI LACRGFSGHN LEYLLRLADF MQLCGPQAQD Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%
CHAC1 Polyclonal antibody specifically detects CHAC1 in Human samples. It is validated for Western Blot.Specifications
CHAC1 | |
Polyclonal | |
Unconjugated | |
CHAC1 | |
1810008K03Rik; Blocks Notch protein; botch; cation transport regulator homolog 1; cation transport regulator-like protein 1; ChaC glutathione specific gamma-glutamylcyclotransferase 1; ChaC glutathione-specific gamma-glutamylcyclotransferase 1; ChaC, cation transport regulator 1; ChaC, cation transport regulator homolog 1; ChaC, cation transport regulator-like 1; Chac1; Gamma-GCG 1; gamma-GCT acting on glutathione homolog 1; glutathione-specific gamma-glutamylcyclotransferase 1; Neuroprotective protein 7; Npg7; RGD1307153 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
79094 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9BUX1 | |
CHAC1 | |
Synthetic peptide directed towards the C-terminal of human CHAC1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |