missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chaetocin, Tocris Bioscience™
Click to view available options
Quantity:
1 mg
Description
Histone methyltransferase SUV39H1 inhibitor (IC50 = 0.8 μM). Induces apoptosis in myeloma cell lines in vitro; exhibits antiproliferative activity in a mouse myeloma model in vivo.
Specifications
Specifications
CAS | 28097-03-2 |
Molecular Formula | C30H28N6O6S4 |
Quantity | 1 mg |
Synonym | chaetocin |
InChI Key | PZPPOCZWRGNKIR-YVNRJRTHSA-N |
SMILES | CN1C(=O)C23CC4(C(N2C(=O)C1(SS3)CO)NC5=CC=CC=C54)C67CC89C(=O)N(C(C(=O)N8C6NC1=CC=CC=C71)(SS9)CO)C |
Molecular Weight (g/mol) | 696.83 |
PubChem CID | 131852036 |
Formula Weight | 696.84 |
Percent Purity | >99% |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'