missing translation for 'onlineSavingsMsg'
Learn More

Charcoal, Technical, ActIVated for Gas Adsorption, 10-18 Mesh, Granular, Fisher Chemical™

Chemical Identifiers

CAS 7440-44-0
Molecular Formula C
MDL Number 133992
View More Specs

Description

Description

Specifications

Chemical Identifiers

7440-44-0
C
133992

Specifications

7440-44-0
Black
C
12g/mol
3652°C
Plastic powder jar
133992
Solid
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Activated charcoal

SDS
Documents

Documents

Product Certifications