missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Charcoal, Technical, Decolourising Powder, ActIVated, Fisher Chemical™
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Specifications
Melting Point | 3652°C |
Color | Black |
Packaging | Plastic powder jar |
Quantity | 1 kg |
Formula Weight | 12g/mol |
Physical Form | Powder |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS02_Extremely_Flammable.png)
Product Identifier
- Charcoal, Activated
- Warning
- Self-heating substance Category 2
- H252-Self-heating in large quantities; may catch fire.
- P235+P410-Keep cool. Protect from sunlight.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'