missing translation for 'onlineSavingsMsg'
Learn More
Charcoal, Technical, Decolourising Powder, ActIVated, Fisher Chemical™ Product Code.: 11478103

Charcoal, Technical, Decolourising Powder, ActIVated, Fisher Chemical™

Product Code. 11478103
500 g, Plastic powder jar
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
This item is not returnable. View return policy
TSE/BSE statement is not available for this product
 

Product Code. 11478103

Brand: Fisher Chemical C/4040/53

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
TSE/BSE statement is not available for this product
 

Chemical Identifiers

CAS 7440-44-0
Molecular Formula C
MDL Number 133992

Specifications

Melting Point 3652°C
Color Black
Packaging Plastic powder jar
Quantity 500 g
Formula Weight 12g/mol
Physical Form Powder
compliance-icons
Product Identifier
  • Charcoal, Activated
Signal Word
  • Warning
Hazard Category
  • Self-heating substance Category 2
Hazard Statement
  • H252-Self-heating in large quantities; may catch fire.
Precautionary Statement
  • P235+P410-Keep cool. Protect from sunlight.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • Multiple Manufacturers
    Shop now and save up to 97% on top-quality products for your lab needs.
    Promo Code: 25908

    Expires2025-03-31

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Charcoal, Technical, Decolourising Powder, ActIVated, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.