missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Charcoal, Technical, SLR, Lump, Wood, Fisher Chemical
Click to view available options
1kg, Plastic powder jar
Specifications
Specifications
CAS | 7440-44-0 |
Color | Black |
Packaging | Plastic powder jar |
Molecular Formula | C |
MDL Number | 133992 |
Quantity | 1 kg |
Physical Form | Solid |
Safety and Handling
Product Identifier
- Charcoal, powder, animal
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Charcoal, Technical, SLR, Lump, Wood, Fisher Chemical >
Spot an opportunity for improvement?Share a Content Correction