Learn More
Invitrogen™ Chargeswitch™ Pro Filter Collection Tube
Chargeswitch™ Pro Filter Collection Tube
Brand: Invitrogen™ CS32000
Code : 50
Additional Details : Weight : 0.31000kg
Description
The Chargeswitch™ -Pro Filter Collection Tube is a 50 ml conical tube that will accommodate the centrifuge protocol of Chargeswitch™ Pro Filter Plasmid Midi and Maxi kitsSpecifications
50 mL | |
25 | |
0.01 gal. | |
Conical Tube |
For Research Use Only. Not for use in diagnostic procedures.