Learn More
Eppendorf™ Charging Cradles
Adapt charging stand 4880 000.026 to store and charge a variety of different liquid handling devices. Eppendorf Charging Cradles are designed for use with Eppendorf charger carousels.
Brand: Eppendorf™ 4880601003
This item is not returnable.
View return policy
Description
- Charger shell for charger carousel (4880 000.026)
- Adapter for Multipette
Specifications
Charger carousel (4880 000.026) | |
1 Each |
Product Content Correction
Product Title
Eppendorf™ Charging Cradles > Charging Cradle, For Charging Carousel Series 4880 (X) Stream
Spot an opportunity for improvement?Share a Content Correction