missing translation for 'onlineSavingsMsg'
Learn More

LMS™ Chart Recorder Paper for Cooled Incubator

Product Code. 12813456
Click to view available options
Product Type:
Roll LMS CRP28 (Tear-off)
Roll LMS CRP29 (Re-roll)
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12813456

Brand: LMS™ CRP29

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

LMS™ Cooled Incubators come with a wide range of factory-fitted optional accessories and other extras to cater for individual requirements and even more operational flexibility.

  • Roll of chart recorder paper
  • Available in tear-off or re-roll formats
  • Accessory for use with the optional  LMS™ cooled incubator chart recorder
  • Sufficient for thirty-day recording

Specifications

Product Type Roll LMS CRP29 (Re-roll)
Description Recorder paper
For Use With (Application) Chart Recorder(Roll)
For Use With (Equipment) LMS™ Chart Recorder
Quantity 1 Roll
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.