missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LMS™ Chart Recorder Paper for Cooled Incubator
LMS™ Cooled Incubators come with a wide range of factory-fitted optional accessories and other extras to cater for individual requirements and even more operational flexibility.
£62.00 - £90.00
Specifications
Description | Recorder paper |
---|---|
For Use With (Equipment) | LMS™ Chart Recorder |
Quantity | 1 Roll |
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
12522597
|
LMS™
CRP28 |
Roll LMS CRP28 (Tear-off) |
£90.00
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||||
12813456
|
LMS™
CRP29 |
Roll LMS CRP29 (Re-roll) |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Roll of chart recorder paper
- Available in tear-off or re-roll formats
- Accessory for use with the optional LMS™ cooled incubator chart recorder
- Sufficient for thirty-day recording
Specifications
Recorder paper | |
LMS™ Chart Recorder | |
1 Roll |