Learn More
Thermo Scientific™ Chart Recorders for Ultra-Low Temperature Freezers
Record seven days of temperature performance at temperatures to -150° C.
£64.88 - £1789.81
Product Code | Brand | Description | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | For Use With (Equipment) | Price | Quantity & Availability | ||||
11880573
|
Thermo Scientific™
17020 |
Ink Chart Paper (pack of 50) | HERAfreeze™ HFU-B, Forma™ 900, Revco™ ExF/DxF, TSE/TSD Series |
£99.00
Pack of 50 |
In Stock
Log in to see stock. |
||||
13574930
|
Thermo Scientific™
180000 |
Inkless Chart Paper (pack of 50) | Forma™ 900, HERAfreeze™ HFU-B, Revco™ ExF/DxF, TSE/TSD Series |
£120.47
Pack of 50 |
Estimated Shipment: 14-10-2024
Log in to see stock. |
||||
10298002
|
Thermo Scientific™
201240 |
Ink Chart Recorder, Field installed | TSE/TSD Series, Forma™ 900, HERAfreeze™ HFU-B, Revco™ ExF/DxF |
£1660.74
Each |
Estimated Shipment: 01-11-2024
Log in to see stock. |
||||
10715392
|
Thermo Scientific™
201241 |
Ink Chart Recorder, Factory installed | TSE/TSD Series, Forma™ 900, HERAfreeze™ HFU-B, Revco™ ExF/DxF |
£1307.94
Each |
Estimated Shipment: 17-10-2024
Log in to see stock. |
||||
10606013
|
Thermo Scientific™
201248 |
Inkless Chart Recorder, Field Installed | HERAfreeze™ HFU-B, Forma™ 900, Revco™ ExF/DxF, TSE/TSD Series |
£1359.57
Each |
In Stock
Log in to see stock. |
||||
11983927
|
Thermo Scientific™
201249 |
Inkless Chart Recorder, Factory Installed | Forma™ 900, HERAfreeze™ HFU-B, Revco™ ExF/DxF, TSE/TSD Series |
£1368.18
Each |
Estimated Shipment: 18-10-2024
Log in to see stock. |
||||
10553984
|
Thermo Scientific™
245232 |
Felt-tip pen, blue | Forma™ 900, HERAfreeze™ HFU-B, Revco™ ExF/DxF, TSE/TSD Series |
£64.88
Each |
Estimated Shipment: 17-10-2024
Log in to see stock. |
||||
11336351
|
Thermo Scientific™
398223 |
Ink Chart Recorder, Field installed | Revco CxF Series (except ULT350 and ULT390) |
£1505.85
Each |
Estimated Shipment: 18-10-2024
Log in to see stock. |
||||
11346351
|
Thermo Scientific™
398224 |
Ink Chart Recorder, Factory installed | Revco CxF 390, 350 |
£1368.18
Each |
Estimated Shipment: 18-10-2024
Log in to see stock. |
||||
11356351
|
Thermo Scientific™
398225 |
Ink Chart Recorder, Field installed | Revco CxF 390, 350 |
£1505.85
Each |
Estimated Shipment: 18-10-2024
Log in to see stock. |
||||
10601151
|
Thermo Scientific™
6185 |
Inkless Chart Paper (pack of 50) | Thermo Scientific™ TSU Series, HERAfreeze™ HFU-T, Revco™ UxF |
£86.05
Each |
In Stock
Log in to see stock. |
||||
16318158
|
Thermo Scientific™
6383-6V |
Inkless Chart Recorder, Field Installed | Forma™ 88300, HERAfreeze™ HFU300T, Revco™ UxF300 |
£1712.37
Each |
Estimated Shipment: 25-09-2024
Log in to see stock. |
||||
16357468
|
Thermo Scientific™
6383-6W |
Inkless Chart Recorder, Field Installed | Forma™ 88300, HERAfreeze™ HFU300T, Revco™ UxF300 |
£1712.37
Each |
Estimated Shipment: 09-10-2024
Log in to see stock. |
||||
11366381
|
Thermo Scientific™
CR4567 |
Inkless Chart Recorder, Factory installed | TSX Series, Forma FDE, HERAfreeze HDE, Revco RDE and TDE Series (400 box capacity and larger) |
£1789.81
Each |
Estimated Shipment: 18-10-2024
Log in to see stock. |
||||
11336391
|
Thermo Scientific™
FCR400R |
Inkless Chart Recorder, Field Installed | TSU400, Revco™ UxF400 |
£1712.37
Each |
Estimated Shipment: 18-10-2024
Log in to see stock. |
||||
11346391
|
Thermo Scientific™
FCR567FTH |
Inkless Chart Recorder, Field Installed | Forma™ 88500, Forma™ 88600, Forma™ 88700, TSU500, TSU600 |
£1712.37
Each |
Estimated Shipment: 18-10-2024
Log in to see stock. |
||||
11356391
|
Thermo Scientific™
FCR567R |
Inkless Chart Recorder, Field Installed | HERAfreeze™ HFU700T, HERAfreeze™ HFU600T, Revco™ UxF500, Revco™ UxF600, Revco™ UxF700 |
£1712.37
Each |
Estimated Shipment: 23-10-2024
Log in to see stock. |
||||
11993907
|
Thermo Scientific™
FCR400FT |
Inkless Chart Recorder, Field Installed | Forma™ 88400, TSU400 |
£1712.37
Each |
Estimated Shipment: 18-10-2024
Log in to see stock. |
||||
15389615
|
Thermo Scientific™
FCR567TSX |
Inkless Chart Recorder, Field Installed | TSX Series |
£1712.37
Each |
Estimated Shipment: 18-10-2024
Log in to see stock. |
||||
15369615
|
Thermo Scientific™
FCRP567TSX |
Ink Chart Recorder, Field Installed | TSX Series |
£1712.37
Each |
Estimated Shipment: 18-10-2024
Log in to see stock. |
||||
15788527
|
Thermo Scientific™
FCR400TSXLE |
Inkless Chart Recorder, Field Installed | TSX400 ultra-low freezers |
£1712.37
Each |
Estimated Shipment: 06-11-2024
Log in to see stock. |
||||
15708537
|
Thermo Scientific™
FCR500TSXLE |
Inkless Chart Recorder, Field Installed | TSX500 ultra-low freezers |
£1712.37
Each |
Estimated Shipment: 06-11-2024
Log in to see stock. |
||||
15728537
|
Thermo Scientific™
FCR600TSXLE |
Inkless Chart Recorder, Field Installed | TSX600 ultra-low freezers |
£1712.37
Each |
Estimated Shipment: 18-09-2024
Log in to see stock. |
||||
15738537
|
Thermo Scientific™
FCR700TSXLE |
Inkless Chart Recorder, Field Installed | TSX700 ultra-low freezers |
£1712.37
Each |
Estimated Shipment: 06-11-2024
Log in to see stock. |
||||
15758537
|
Thermo Scientific™
FCRP400TSXLE |
Ink Chart Recorder, Field Installed | TSX400 ultra-low freezers |
£1712.37
Each |
Estimated Shipment: 06-11-2024
Log in to see stock. |
||||
15778537
|
Thermo Scientific™
FCRP500TSXLE |
Ink Chart Recorder, Field Installed | TSX500 ultra-low freezers |
£1712.37
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
||||
15798537
|
Thermo Scientific™
FCRP600TSXLE |
Ink Chart Recorder, Field Installed | TSX600 ultra-low freezers |
£1712.37
Each |
Estimated Shipment: 06-11-2024
Log in to see stock. |
||||
15708547
|
Thermo Scientific™
FCRP700TSXLE |
Ink Chart Recorder, Field Installed | TSX700 ultra-low freezers |
£1712.37
Each |
Estimated Shipment: 06-11-2024
Log in to see stock. |
||||
15778527
|
Thermo Scientific™
FCR400LE |
Inkless Chart Recorder, Field Installed | Revco RLE, Forma 89000, TLE Series, HLE Series (400 models only) |
£1712.37
Each |
Estimated Shipment: 06-11-2024
Log in to see stock. |
||||
15798527
|
Thermo Scientific™
FCR500LE |
Inkless Chart Recorder, Field Installed | Revco RDE, Forma FDE, TDE Series, HDE Series (500 box capacity models only) |
£1712.37
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
||||
15718537
|
Thermo Scientific™
FCR600LE |
Inkless Chart Recorder, Field Installed | Revco RDE, Forma FDE, TDE Series, HDE Series (600 box capacity models only) |
£1712.37
Each |
Estimated Shipment: 17-09-2024
Log in to see stock. |
||||
15748537
|
Thermo Scientific™
FCRP400LE |
Ink Chart Recorder, Field Installed | Revco RLE, Forma 89000, TLE Series, HLE Series (400 models only) |
£1712.37
Each |
Estimated Shipment: 06-11-2024
Log in to see stock. |
||||
15768537
|
Thermo Scientific™
FCRP500LE |
Ink Chart Recorder, Field Installed | Revco RLE, Forma 89000, TLE Series, HLE Series (500 models only) |
£1712.37
Each |
In Stock
Log in to see stock. |
||||
15788537
|
Thermo Scientific™
FCRP600LE |
Ink Chart Recorder, Field Installed | Revco RLE, Forma 89000, TLE Series, HERAfreeze HLE 600-box freezers |
£1712.37
Each |
Estimated Shipment: 06-11-2024
Log in to see stock. |
||||
17888000
|
Thermo Scientific™
332166G01 |
Inkless Chart Recorder, Factory Installed | TDE Series Chest Freezers |
£1471.43
Each |
Estimated Shipment: 18-09-2024
Log in to see stock. |
||||
17898000
|
Thermo Scientific™
332166G02 |
Ink Chart Recorder, Factory installed | TDE Series Chest Freezers |
£1471.43
Each |
Estimated Shipment: 18-09-2024
Log in to see stock. |
||||
17818020
|
Thermo Scientific™
332791G01 |
Inkless Chart Recorder, Factory Installed | TDE Series Chest Freezers |
£1471.43
Each |
Estimated Shipment: 18-09-2024
Log in to see stock. |
||||
17828020
|
Thermo Scientific™
332791G02 |
Ink Chart Recorder, Factory installed | TDE Series Chest Freezers |
£1471.43
Each |
Estimated Shipment: 18-09-2024
Log in to see stock. |
||||
Description
Record seven days of temperature performance at temperatures to -150° C using Thermo Scientific™ Chart Recorders for Ultra-Low Temperature Freezers. Recorders use 6″ (15cm) chart paper and are available as factory or field installed.
Features:- Seven-day recorder
- Functions to -150°C
- Field or factory installed
Ordering information:
- Field-installed units must be installed by a qualified professional
Specifications
Recorder |