Learn More
CHAT Mouse anti-Human, Unconjugated, Clone: 1F7A9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66816-1-IG-150UL
This item is not returnable.
View return policy
Description
The CHAT antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CHAT. This antibody recognizes human, mouse, rat antigen. The CHAT antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
CHAT | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, ELISA | |
1F7A9 | |
Unconjugated | |
150 μL | |
1103 | |
IgG2b |