missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHD3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | CHD3 |
---|---|
Concentration | 0.35 mg/mL |
Applications | Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16803925
|
Proteintech
25335-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16893915
|
Proteintech
25335-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a member of the CHD family of proteins which are characterized by the presence of chromo domains and SNF2-related helicase/ATPase domains. This protein is one of the components of a histone deacetylase complex referred to as the Mi-2/NuRD complex which participates in the remodeling of chromatin by deacetylating histones. Chromatin remodeling is essential for many processes including transcription. Autoantibodies against this protein are found in a subset of patients with dermatomyositis. Three alternatively spliced transcripts encoding different isoforms have been described.Specifications
CHD3 | |
Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human | |
Q12873 | |
1107 | |
Peptide | |
Primary | |
-20°C | |
CHD3 |
0.35 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ATP dependent helicase CHD3, CHD 3, CHD3, hZFH, Mi 2a, Mi2 ALPHA, ZFH, Zinc finger helicase | |
CHD3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |