Learn More
Check Standards, Palintest™
Click to view available options
Description:
Check Standards Palintest
Check Standards for Ammonia Meter 1000
Check Standards for Bromometer Duo
Check Standards for Chloridiox Meter 1000
Ozone meter 1000 Check Standards With Test Certificate M2 version
Description
- Traceable reference materials defined in terms of % transmittance (%T) at a particular wavelength.
- Ideal in confirming that a Palintest™ photometer is conforming to specification.
- Each standard enables users to check each of the individual wavelengths in the photometer.
Specifications
Description | Check Standards for Ammonia Meter 1000 |
Type | Check Standard |
For Use With (Equipment) | Ammonia Meter 1000 |