Learn More
Lovibond™ Checkit™ Comparator Single Parameter Test Kit
Aqualytic™ Checkit™ Comparator Single Parameter Test Kit includes all materials and operating instructions needed for mobile and static water analysis of a single parameter and comes in a sturdy and handy plastic case.
Brand:  Lovibond™ 147110
 View more versions of this product
Product Code. 10550663
  • £54.75 / Each
Estimated Shipment: 17-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Aqualytic™ Checkit™ Comparator Single Parameter Test Kit includes all materials and operating instructions needed for mobile and static water analysis of a single parameter and comes in a sturdy and handy plastic case.
  • Compact and lightweight
  • Provides the basis for a comprehensive and easy-to-use analysis system
  • Comparator Discs contain a continuous color scale for exact color matching between standard and sample
Specifications
Specifications
pH
Comparator apparatus, color comparison discs, cells, stirring rod, reagents, instructions
5.2 to 6.8mg/L
For Both Mobile and Static Water Analysis
1 Each
Checkit™ Comparator Single Parameter Test Kit