missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ CHECKIT™ Comparator Test Kits: Chlorine-Free/Total
Accurate test kits for routine water analysis
Brand: Lovibond™ 147000
Code : A0
Additional Details : Weight : 0.25000kg
Includes:
- Aqua Comparator Housing (D55 for L147000)
- Aqua Comparator Disc
- DPD No. 1 Rapid Tablets
- DPD No. 3 Rapid Tablets
- 2 x 13.5mm Sample Cells
- Tablet Crusher
- Instructions
- Carrying case
Description
Accurate test kits for routine water analysis
- Easy to use: add reagent to sample cell, rotate disc until color matches sample, and read concentration value
Specifications
±5% | |
Aqua comparator™ housing, aqua comparator™ disc D55, DPD No. 1 tablets, DPD No. 3 tablets, 2 x 13.5mm sample cells, tablet crusher, instructions, carrying case | |
0.02 to 0.3mg/L |
Routine water analysis | |
100 | |
Chlorine Free and Total Test Kit |