Learn More
Hanna Instruments™ Checktemp Digital Thermometers
Digital thermometer with stainless steel penetration probe that delivers high accuracy temperature measurements over a wide temperature range without worrying about breakage or condensation.
Brand:  Hanna Instruments™ HI98501
 View more versions of this product
Product Code. 15350684
  • £152.00 / Each
Explore more special offers
Description
Description
  • A digital thermometer with stainless steel penetration probe. It delivers high accuracy temperature measurements over a wide temperature range without worrying about breakage or condensation
  • Checktemp™ is the ideal instrument for measuring temperature according to HACCP requirements
  • ±0.2°C (±0.5°F) - Accuracy
  • Most accurate temperature tester in the world
  • CAL Check™ - Automatically verifies calibration at startup
  • °C/°F Status – User selectable
  • Large display with wide environmental temperature range and viewing angle.
  • IP 65 water resistant protection
  • HACCP Compatible - Use as a tool for control in HACCP analysis
  • AISI 316 stainless steel penetration Probe
  • 3000 hours of battery life (Continuous use)
Specifications
Specifications
AISI 316, HACCP Compatible
Education, food (storage, transportation, manufacturing, cooking), laboratory
Fixed
51.3 g
150°C
-50°C to +150°C
±0.3°C
3.6 mm
Probe
0.1°C
Digital Thermometer
-50°C
Yes
Digital