missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ Checktemp Digital Thermometers

Digital thermometer with stainless steel penetration probe that delivers high accuracy temperature measurements over a wide temperature range without worrying about breakage or condensation.

Brand:  Hanna Instruments™ HI98509

 View more versions of this product

Product Code. 15379353

  • £99.05 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Unique calibration check feature
  • Unique high accuracy of ±0.3°C between the critical range of -20°C to 90°C
  • Choice of models with fixed probe or probe on 1m cable
    • A high-accuracy thermometer with a 1 m (3.3’) flexible, silicone cable connecting the meter and the stainless steel probe. 
    • CAL Check™ - Automatically verifies calibration at startup
    • Silicone probe cable - 3.3’ silicone cable maintains flexibility and performance
    • °C/°F Status – User selectable
    • Large display with wide environmental temperature range and viewing angle.
    • IP 65 water resistant protection
    • HACCP Compatible - Use as a tool for control in HACCP analysis
    • AISI 316 stainless steel penetration Probe
    • Auto-Off (select from 8 min., 60 min., or turn the feature off)
    • Recommended use in education, food (storage, transportation, manufacturing, cooking), laboratory
Specifications

Specifications

AISI 316, HACCP Compatible
Education, food (storage, transportation, manufacturing, cooking), laboratory
Probe
110 mm
80 g
150°C
-50°C to +150°C
±0.3°C
3 mm
106 mm
1 m
0.1°C
Digital Thermometer
-50°C
Yes
Digital
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.