Learn More
Fisherbrand™ Cheesecloth Wipes
Use these wipes for wiping cuvettes, microscopes, glassware, syringes, pipettes and pH electrodes. Fisherbrand™ Cheesecloth Wipes absorb 6 times their weight of solvents and aqueous solutions.
Brand: Fisherbrand™ 2050
This item is not returnable.
View return policy
Description
- Meets all cleanliness specifications of USP (United States Pharmacopoeia) and Federal Specification CCC-G-101c
- Certified to be 100% pure cotton of reagent-grade quality and purity
- Can be used as a filtering material due to their inertness to harsh solvent
Specifications
Wipes | |
Cheesecloth | |
60 Pk. |