Learn More
Chemical Binder Spillage Treatment, Pure, Fisher Chemical™
Universally applicable chemical binder suitable for use with all chemical spills.
Brand: Fisher Chemical S/7435/53
79.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Code : 24
Additional Details : CAS Number : 25322-68-3 Weight : 0.50000kg
Description
Universally applicable chemical binder suitable for use with all chemical spills.
- Binds with liquids with a volume up to 75 times its own weight
- Does not eliminate the hazards associated with a spillage and the residue must be handled and disposed of according to the substance bound
Specifications
Green | |
500 g |