missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JULABO™ Refrigerators for Chemicals, KRC Series
Julabo™ Refrigerators for Chemicals is used for storing and cooling of chemicals and hazardous substances. The spark free interior prevents damage caused by spilled or evaporating chemicals.
£2097.99
Specifications
Description | Refrigerator for chemicals |
---|---|
Temperature Range | -2°C to +12°C |
Temperature Resolution | 0.1°C |
Temperature Sensor | PTC |
Temperature Stability | ±1°C |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10739844
|
JULABO™
8800705 |
68 L |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- With spark free interior
- Storing and cooling of hazardous substances
- Self-protecting control circuit
- Digital temperature display (LED)
- Overload protection for cooling compressor with test button
- Cut-off in case of disturbance with optical alarm signal
- Working and safety sensors are protected against short circuits and disconnection
Specifications
Refrigerator for chemicals | |
0.1°C | |
±1°C | |
Designed for storing and cooling hazardous substances | |
230 V 50 Hz |
-2°C to +12°C | |
PTC | |
LED | |
230 V | |
50 Hz |