Learn More
Bochem™ Chemical Scoop
Manufactured from 18/10 stainless steel. Bochem™ Chemical Scoop has a round barrel.
Brand:  Bochem™ 8803
Code : A0
Additional Details : Weight : 0.40500kg
 View more versions of this product
Product Code. 10653161
  • £32.15 / Each
In Stock
to see stock.
Add to basket
Description
Description
  • Round barrel
  • High sides
  • 18/10 stainless steel
Specifications
Specifications
Chemical Scoop
1500 mL
350 mm
Stainless Steel (18/10)
120 mm