Learn More
Fisherbrand™ Chemical Sorbent Particulate
Absorbs quickly, thanks to its fast wicking action. Encapsulates spills on contact.
Brand:  Fisherbrand™ FBEU27
375.33 GBP valid until 2025-04-30
Use promo code "24163" to get your promotional price.
Product Code. 15873720
  • £419.12 / Each
Estimated Shipment: 08-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Ideal for oil clean up on open water and land applications
  • Fibrous sorbent for non-aggressive spills of oil based or solvent based liquids
  • Compressed bale pack
  • Loose fibrous peat moss structure
Spillage, absorbent, spill, sorbent
Specifications
Specifications
Brown
Large Fiber Sphagnum Peat Moss
Bale
12.3 kg
100 L
Chemical Sorbent
Fibrous Sorbent for non-aggressive spills of Oil Based or Solvent Based Liquids.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Chemical Sorbent Particulate > Chemical Sorbent Particulate