missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Chemical Sorbent Pillow

A chemical sorbent pillow filled with green polypropylene that absorbs chemicals and hazardous liquids. Ideal for absorbing spills and leaking fluids in tight spaces or under persistent drip sources.

Brand:  Fisherbrand™ FBNHAZ99

263.15 GBP valid until 2025-04-30
Use promo code "24163" to get your promotional price.


Product Code. 15813730

  • £286.70 / Pack of 32
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Green polypropylene filling
  • Ideal for spills and leaking fluids, or under persistent drip sources
  • Perfect for use in small or hard to reach areas

Spillage, absorbent, spill, sorbent

Specifications

Specifications

Hydrophilic, Oleophilic
Chemical absorbent pillow
Meltblown, Spunbond
Under persistent drip sources, in tight spaces, for large spills once contained. Water-base, oil-base, or solvent-base liquids
230 x 230 mm
230 mm
4.5 kg
Green
42 L
Chemical Absorbent
32/Pk.
230 mm
230 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

  • Fisherbrand
    Get up to 60% off select Fisherbrand products
    Promo Code: 24163

    Expires2025-04-30

    New Offer

    *Discount will apply to qualifying orders received by Fisher Scientific between 01/04/2025 and 30/04/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.

    Terms & Conditions Save Now
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Chemical Sorbent Pillow > Chemical Sorbent Pillow

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.