Learn More
Fisherbrand™ Chemical Sorbent Socs
Chemical sorbent socs absorb chemicals and hazardous liquids. Strong wicking action, absorption begins instantly with no need to turn the soc.
Brand: Fisherbrand™
115.62 GBP valid until 2024-07-31
Use promo code "21674" to get your promotional price.
Additional Details : Weight : 3.61800kg
Description
- Strong wicking action: absorption begins immediately after contact with spilled liquid
- No need to turn the soc
- Flexible design allows soc to fit around any machine, corner or curve
- Ideal for containing and absorbing leaks and spills
Spillage, absorbent, spill, sorbent
Specifications
Hydrophilic, Oleophilic | |
Chemical sorbent soc | |
Spunbond Meltblown Polypropylene | |
Chemical resistant, universal sorbent sock for spills of water base, oil base, or solvent base liquids | |
1.2 m x 76 mm | |
76 mm |
Green | |
43 L | |
Chemical Sorbent | |
12/Cs. | |
1.2 m | |
5.3 kg |