missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Chemical Spill Kit, Absorption Capacity 16L
This economy portable spill kit comes in a high visibility, water-resistant, yellow PVC bag with handles. Designed for small spills of chemicals and hazardous liquids. Ideal for a small storage areas, such as behind or under a seat thanks to its compact size.
Brand: Fisherbrand™ FBSKHPPN
90.76 GBP valid until 2025-03-31
Use promo code "24258" to get your promotional price.
Description
- Economy chemical spill kit
- Portable and easy to store
- High visibility, water-resistant bag keeps sorbents dry
- Light weight, ideal for carriers transporting small quantities of liquids
Spillage, absorbent, spill, sorbent
Specifications
Yellow | |
Economy Chemical Spill Kit | |
10 x pads (500 x 400 mm), 2 x HAZ412 SOCs (76 mm dia. x 1220 mm), 1 pair of gloves, 1 plastic disposal bag and instructions | |
500 x 400 x 110 mm | |
400 mm | |
1.8 kg |
16 L | |
Emergency Kit for Chemical Spill | |
1 Kit | |
500 mm | |
110 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Chemical Spill Kit, Absorption Capacity 16L > Chemical Spill Kit
Spot an opportunity for improvement?Share a Content Correction