missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Chemical Spill Kit, Absorption Capacity 37L
Description
- Portable spill kit
- Designed for small spillages
- Durable see-through bag with shoulder strap - easy to check spill response supplies
- Quick and easy access ensures a quick response, thanks to a zipper opening up the entire front of the bag
Spillage, absorbent, spill, sorbent
Specifications
Saturated, Medium Viscosity | |
37 L | |
Emergency response spill kit (Hydrophilic + Oleophilic + Hazardous Chemicals) | |
1 Kit | |
400 mm |
Yellow | |
Portable Chemical Spill Kit | |
25 pads (500 x 400 mm), 4 SOCs (1220 mm), 1 yellow disposal bag, 1 pair of gloves | |
500 mm | |
200 mm |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Chemical Spill Kit, Absorption Capacity 37L > Chemical Spill Kit, Absorption Capacity 37L
Spot an opportunity for improvement?Share a Content Correction