missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ Chemical Spoon, Spatula Handle, Nickel

Dual-purpose design provides versatility. Bochem™ Chemical Spoon, Spatula Handle, Nickel is a long-handled spoon with spoon and spatula ends.

Brand:  Bochem™ 3431

Product Code. 10324031

  • £64.05 / Each
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Spoon and spatula ends.
  • Long handle.
  • Resistant Nickel (99.5%) construction.
Specifications

Specifications

Spoon Spatula
22 mm
150 mm
32 mm
Nickel (99.5%)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers