Learn More
Hanna Instruments™ Chemical Test Kit
Includes all of the reagents and equipment necessary to perform manual water testing. Hanna Instruments™ Chemical Test Kit features pre-made, pre-measured reagents for fast, accurate determinations with high-resolution readings based on accepted titrations methods.
Brand:  Hanna Instruments™ HI3833
 View more versions of this product
Product Code. 10634155
  • £42.50 / Each
Explore more special offers
Description
Description
These chemical test kits use high quality reagents that are manufactured in Hanna's state-of-the-art facility and are clearly marked with the lot number and expiration date on each packet/bottle for traceability.
  • Complete setup: All required materials are included, such as beakers, syringes, and premeasured reagent bottles and packets
  • High resolution readings for fast, accurate determinations
  • Replacement reagents available: No need to buy a new kit when reagents are exhausted
Specifications
Specifications
Ascorbic acid
Water Samples
50
0 to 5mg/L
Phosphate Test kit
Phosphate determination
20mL plastic beaker, color comparison cube, and 50 packets phosphate reagent
1 Kit
Titration
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products