missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hanna Instruments™ Chemical Test Kit
Includes all of the reagents and equipment necessary to perform manual water testing. Hanna Instruments™ Chemical Test Kit features pre-made, pre-measured reagents for fast, accurate determinations with high-resolution readings based on accepted titrations methods.
Brand: Hanna Instruments™ HI3822
View more versions of this product
This item is not returnable.
View return policy
Description
These chemical test kits use high quality reagents that are manufactured in Hanna's state-of-the-art facility and are clearly marked with the lot number and expiration date on each packet/bottle for traceability.
- Complete setup: All required materials are included, such as beakers, syringes, and premeasured reagent bottles and packets
- High resolution readings for fast, accurate determinations
- Replacement reagents available: No need to buy a new kit when reagents are exhausted
Specifications
Sulfite determination | |
30mL sulfamic acid solution, 30mL EDTA reagent, 15mL sulfuric acid solution, 10mL starch indicator, 120mL titrant solution, 20mL calibrated vessel, 50mL calibrated vessel, and calibrated syringe with tip | |
1 Kit | |
Two ranges | |
Sulfite Test Kit |
Water samples | |
110 | |
0 to 20mg/L and 0 to 200mg/L | |
Iodometric |