Learn More
Thermo Scientific™ Cheminert™ Sample injector loops, PAEK, 5μL
Use Thermo Scientific™ Cheminert™ Model C2 Microbore Injectors as injectors or switching valves.
Brand: Thermo Scientific™ CZSL5PK
This item is not returnable.
View return policy
Description
Use Thermo Scientific™ Cheminert™ Model C2 Microbore Injectors as injectors or switching valves. It can be actuated manually or with a microelectronic actuator. With 1/16 in. fittings and 0.010 in. ports, it is available in six-port or ten-port configurations and includes a 5μL sample loop.
- 1/16″ fittings
- 0.010″ ports
- Available in 6-port or 10-port configurations
- Available with manual or microelectric actuator
Specifications
5 μL | |
Sample Injector Loop | |
PEEK |