Learn More
Thermo Scientific™ Cheminert™ Sample injector loops, stainless steel, 5μL
Product Code. 11593960 Shop All Thermo Scientific Products
Click to view available options
Volume (Metric) Sample:
5 μL
Unit Size:
Each
Product Code. 11593960
Brand: Thermo Scientific™ CSL5
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use Thermo Scientific™ Cheminert™ Model C2 Microbore Injectors as injectors or switching valves.
Use Thermo Scientific™ Cheminert™ Model C2 Microbore Injectors as injectors or switching valves. It can be actuated manually or with a microelectronic actuator. With 1/16 in. fittings and 0.010 in. ports, it is available in six-port or ten-port configurations and includes a 5μL sample loop.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) Sample 5 μL
Type Sample Injector Loop
Stator Material Stainless Steel