missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ ChemTek™ 38-520 Series Butyl Polymer Immersion Gloves

Product Code. 12659711 Shop All Ansell Products
Click to view available options
Size:
7
8
9
10
11
Unit Size:
1 pair

Product Code. 12659711

Brand: Ansell™ 38520080

Only null left
Add to basket
Add to basket

Provides the best resistance to the most aggressive chemicals without compromising dexterity or comfort. ChemTekTM gloves are surprisingly flexible and provide excellent dexterity.

  • Smooth finish in black
  • Easy donning and good grip
  • Maximum protection from hazardous chemicals
  • Surprisingly flexible and provide excellent dexterity
  • Designed for comfort with a natural, curved ergonomic shape and soft feel
  • Absence of latex proteins means there's no risk of latex allergies
  • Ideal for the chemical industry, oil and petrol refining, printing industry, automotive industry, mining and aerospace
TRUSTED_SUSTAINABILITY

Specifications

Back Material Butyl
Cuff Material Butyl
Applications Industrial
Coating Material Butyl Polymer
Color Black
Laboratory/Cleanroom Use Yes
Latex Free Yes
Material Butyl Polymer
Product Type Immersion Gloves
Size 8
Thickness (English) 20 mil
Thickness (Metric) 0.0268 in.
Glove Cuff Color Black
Glove Cuff Style Beaded/Rolled
Hem Color Black
Inspected for Medical Use Yes
Packaging Type Individually wrapped bags with insert
Quantity 1 Pair
Resistant To Highly chemical resistance
Series 38-520
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.