missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Saint Gobain Life Sciences Chemware™ PFA Jar with Cap
Click to view available options
Capacity:
1000 mL (33.8 oz.)
120 mL (4.1 oz.)
2000 mL (67.6 oz.)
360 mL (12.2 oz.)
480 mL (16.2 oz.)
Description
- Chemware™ PFA jars store solutions, paste and bulk materials with no concern about leaching, corrosion, contamination or evaporation.
- They are resistant to corrosive liquids, acids, solvents, sensitive reagents, and staining solutions.
- PFA jars feature a large mouth for easy access and non-stick surface for easy cleaning.
- There is an area for identification marking on side of the jars.
- Stackable, these jars have a knurled top for secure grip to facilitate easy opening and closing.
Specifications
Capacity | 120 mL (4.1 oz.) |
Capacity (Metric) | 120 mL |
Capacity (English) | 4.1 oz. |
Diameter (English) | 8365008 |
Material | PFA |
Closure Included | Yes |
Mouth | Wide |
Height (Metric) | 53.2 mm |
Product Type | Jar |
Height (English) | 2.1 in. |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'