missing translation for 'onlineSavingsMsg'
Learn More

Saint Gobain Life Sciences Chemware™ Test Tube with Cap

Product Code. 11714518
Click to view available options
Capacity (Metric):
5 mL
20 mL
25 mL
50 mL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11714518

Brand: Saint Gobain Life Sciences D1069690

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Metal-resistant PFA Chemware™ Test Tube with Cap for trace analysis.

  • Chemware™ PFA test tubes are ideal for trace metal/ICP analysis and applications requiring excellent chemical resistance.
  • PFA has the lowest metal ion content and widest temperature range of any fluoropolymer resin.
  • Test tubes are translucent for easy viewing of contents and withstand temperatures up to 260°C (500°F).
  • Can be sterilized repeatedly.
TRUSTED_SUSTAINABILITY

Specifications

Color Translucent
Capacity (Metric) 25 mL
Length (Metric) 185 mm
Material PFA
Shape Cylindrical
Sterility Non-sterile
Graduated No
Product Type Test Tube
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.