missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHES, 0.5M buffer soln, pH 8.5, Thermo Scientific™
Click to view available options
Quantity:
100 mL
250 mL
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Concentration | 0.5 M |
pH | 8.5 |
Physical Form | Liquid |
Quantity | 250 mL |
Chemical Name or Material | CHES buffer soln. |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'